A propos de cet article
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern.
- Diameter 25 mm
- Using #2 (0.17 to 0.25 mm) thickness cover slips
- Unique grid pattern, numbered 1 to 200 squares, each 500 µm width
- Suitable for insertion into most 6-well plates
- Adaptable to most microscope mounting plates
- Facilitates observation of activity growing cells
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern.