CultureWell™ Accessories, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Reusable CultureWell™ gaskets from CultureWell™ Chambered Coverglass and CultureWell™ Chamber Coverglass Inserts are available bulk packaged.
- Gaskets are ideal for forming wells on glass microscope slides or in polystyrene dishes
- Gaskets are non-sterile and may be sterilized by autoclaving, UV or alcohol
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.