Disposable Rinzl Plastic Micro Slides, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Made from optical clear vinyl with a refractive index of glass.
- Ideal for wet mount, urines, and more
- Available standard size 3×1" and packaged 144 per box
- Will not break, scratch, or cut fingers
- Not auto-fluorescent
- Thickness 0.0180 to 0.022" or 0.46 to 0.56 mm
These rinzl plastic micro slides are economical, they can be used and discarded.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.