W PC-620 Ultrasonic Cleaner, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
The PC-620 is offered in 2 frequencies to match cleaning capability to your application. At 40 kHz it is ideal for cleaning pipettes, lab glassware, and other items requiring general cleaning.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.