Specifications
- Description:Paterson Universal Tank & Reel
- Cat. no.:100501-472
- Supplier No.:74342-15
Specifications
About this item
The Super System 4 is simpler to load, faster to fill, and easier to clean than any other tank. It has a large diameter one-piece lid and funnel, which clips into place positively and securely enabling the tank to be filled and emptied quickly. The Paterson 35 mm tank takes a single 35 mm film. The Paterson Universal tank takes two 35 mm films or a single 120/220 film. Supplied complete with one 35mm/120/220 Auto Load Reel, Watertight EVA cap prevents leakage during inversion agitation. Uses less than 300 mL solution for a single 35 mm film.