Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax.
- Available with steel, nylon, fiberglass or brass
- With metal ferrules for improved durability
- Overall length is 41/4" and the brush is 1/8" wide
Packaging: Refills are available and are packaged 12/box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.