Scalpel Handles Disposable, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
These strong, moulded plastic scalpel handles provide a secure palm-fitting grip for post-mortem work, trimming light plaster casts, and similar forceful cutting.
Handle size #6 and #8 Bard-Parker* brand.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.