A propos de cet article
The most convenient, organized and versatile way of storing 75 x 25 mm (3 x 1") microscope slides vertically
SlideFile™ System is a removable tray inside the storage box having 100 numbered slots. In the upright position, one is able to read bar codes without having to remove the slide from the box. For space saving purposes, double the amount of slides simply by storing two slides side-by-side per slot. For maximum storage space-long-term storage-simply remove the SlideFile Tray and line up 400 slides in three rows. Six drain holes incorporated into the tray ensure complete emptying of the liquid when using the slide holder for drying purposes. Two index cards numbered from 1 to 100 are included to allow for slide recording titles. Dimensions: 86 x 248 x 83 mm high (33/8” x 9¾” x 3¼” high).
This impact resistant SlideFile™ Storage System can hold up to 400 slide per unit in just 1720 cm3 (105 cu. in.) and are stackable for space saving. Each SlideFile System includes a slide box and two removable trays. A transparent hinged cover makes it easy to see at a glance the content of the box.