A propos de cet article
This tape dispenser offers you a convenient writing platform, suitable for both left- and right-handed users. The epoxy-coated platform with finger holes and built-in cutting edge, make dispensing and cutting the tape a simple one-handed action. The 3" (76 mm) core fits standard tape rolls up to 1" (25 mm) wide. The weighted base with non-skid bottom assures stability when in use.
- Writing platform
- Weighted base with non-skid pads
- One-handed action
- Weight: 4 lbs (1.8 kg)
- The dispenser accepts series 77010-tapes
The epoxy-coated platform with finger holes and built-in cutting edge, make dispensing and cutting the tape a simple one-handed action. The weighted base with non-skid bottom pads assures stability. The 3" (76mm) core fits standard tape rolls up to 1" (25mm) wide. The weighted base with non-skid bottom assures stability when in use. Weighs 4 lbs (1.8kg). The dispenser accepts series 77010-tapes shown above. The tape is not included.