Vibrating Spatula, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
A toothed wheel is turned by a finger making the spatula blade vibrate.
- Plastic handle
- Stainless steel blade with 'PTFE' coating
This action allows for accurate adding of material.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.