Spécifications
- Description:Trimaide
- Supplier No.:72550
Spécifications
A propos de cet article
A razor blade holder for trimming blocks. Holds all types of razors. It gives you a better hold on the blade while you trim your blocks. The blade slides sideways in the holder allowing use of the entire edge. It protects fingers.