Specifications
- Description:Mold for Long Tissues
- Cat. no.:100491-604
- Supplier No.:70166
Specifications
About this item
This mold accommodates longer pieces of tissues and orient them properly
The tissues lie flat in the well therefore no horizontal trimming is needed. It is ideal for any specimen that needs immediate sectioning (e.g. cell monolayer). The mold has 24 cavities in it.