3M™ Double Sided Tape Scotch 665 or Scotch 666, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Non-conductive transparent tape with adhesive on both sides. Available with or without white paper liner in different widths.
- Good adhesion and cleanliness
- Versions with liner: white paper as liner is slit in the center to facilitate its removal
- 77 mm (3") core
32.9 m (36 yds.) long
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.