3M™ Scotch 616 Lithographer's Tape, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
This ruby-red transparent tape is for use in opaquing, stripping, and edging on film materials. 3" core.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.