Scalpel Handles; Feather®, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Handles for use with different scalpel blades.
- Handle #3; 100mm long, flat with corrugated grip. For use with scalpel blades #10, #11, #12 and #15.
- Handle #4; 105mm long, flat with corrugated grip. For use with scalpel blades #20, #21, #22, #23, and #24.
- Handle #7; 140mm long, slender, contoured with corrugations. For use with scalpel blades #10, #11, #12, and #15
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.