A propos de cet article
Multi purpose glass sample vials suited for research studies. Clear vials are manufactured from Wheaton-33R a low extractable borosilicate glass which protects against changes in the pH and maintains sample integrity. Amber vials are manufactured from Wheaton '320' borosilicate glass and protects light-sensitive samples.
- Economically priced
- Come with lined caps
- Can be sterilized
- Can be stored at low temperatures