Sample Storage Sets; Snap Cap Shell Vial, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom.
Alpha-numerical indexing system allows quick identification and location of stored items.
Ordering information: For vial sizes of 10 and 12 dram please call us for prices and availability.
Delivery: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.