Round Cover Slips, German Glass, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
For neural cell cultures.
- Shown to be more effective with hard to attach cells
- #1 Thickness (0.13 to 0.16 mm)
Packaging: Packed 1 oz. (28 g) per case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.