Round Cover Slips (Bellco Brand), Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Round coverslips with #1 Thickness (0.13 to 0.16 mm).
Packed 1 oz (28g) per case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.