Rectangular and Square Cover Slips (Bellco Glass), Electronic Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
These cover slips provides good quality, reliability, and better performance for all laboratory microscopy needs
- #1 thickness (0.13 to 0.16 mm)
- Packed one ounce (28 g) per case
- Clear white borosilicate glass.
For use with Bellco Leighton Tissue Culture Tubes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.