Spécifications
- Description:Peca Techni-Valve™
- Supplier No.:70849-10
Spécifications
A propos de cet article
Peca Techni-Valve™ for use with Dust-Pro™ Refill cans.
- Smooth bell housing and no mounting clip is required
- Patented bell housing fits the contour of your hand
It is made from solid brass chrome plated is professionally designed with a long nozzle (90 mm (31/2")). The valve can be removed at any time without the loss of pressure.