Poly-L-Lysine Coated and Silane Treated Microscope Slide, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Premium glass slides with a frosted end. These slides improve the adhesion of tissues to the slide. Available pre-coated with either Poly-L-Lysine or Silane (aminoalkysilane).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.