Peel Away Disposable Embedding Molds, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
For plastic or paraffin embedding; no block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.