Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Good for handling infectious materials, inoculating culture media and routine sampling. Available in either Borosilicate or Flint glass, with an outside body diameter of 7 mm.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.