ContactUsLinkComponent
Para-Marker, Electron Microscopy Sciences
Para-Marker, Electron Microscopy Sciences
# de catalogue 100493-278
Minority or Woman-Owned Business Enterprise
Numéro CAS:  
Para-marker
Para-Marker, Electron Microscopy Sciences
# de catalogue 100493-278
Minority or Woman-Owned Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Para-Marker
  • Supplier No.:
    71313-20
Spécifications
A propos de cet article
A wide tipped pen, designed for use with paraffin sections
The pen provides a sticky membrane to the glass slide which not only prevents paraffin sections from falling off but also keeps the sections flat on the slide.
Customers Who Bought This Also Bought