Mixing Spatulas, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Made from polished stainless steel.
They are ideal for applying wax to glass knives, colloidal adhesives to stubs, and for mixing embedding resins.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.