About this item
Bags are made up of porous paper which provides maximum tissue safety during processing.
- Made from porous paper
- Permits proper fluid exchange and drainage
- Available in 25 and 35 mm diameter or 25 and 32 mm sizes
They permit proper fluid exchange and drainage.
To use: simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. Now ready to process as usual.