Paterson Safelight, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Perfect for the small darkroom, or for safety spot lighting individual work areas.
- Freestanding, mounted on the wall, or ceiling
- safe for all black-and-white papers
- 120 V AC; 3-prong cord
DxH: 4x6.25"
Ordering information: The unit comes with a 15 W bulb.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.