Ilford Multigrade IV RC Deluxe Photographic Papers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast. MULTIGRADE IV RC Deluxe has the standard weight of (190g/m2)with a resin coated base.
- Resin Coated – Black & White
- Non-Developer Incorporated
- 12 Contrast Grades with MULTIGRADE Fibers
- Glossy Surface
- Compatible with the Kodak OC Filter
- Dish/Tray Processing
- Developer: Mutigrade, Bromophen or PQ Universal
- Fixer: Ilford Rapid Fixer or Hypam
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.