Disposable Sterile Collecting Bottles, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Disposable polystyrene bottles for the storage of media, buffers and other aqueous solutions.
- Easy grip sides facilitate handling
- Sterile
- Certified non-pyrogenic
- Neck size 45 mm
Plug seal cap provides an airtight seal and helps minimize the risk of contamination.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.