Ideal for filtration of tissue culture media, biological fluids, fixation buffers and more. (not for parenteral use.)
- Vacuum filtration units fit standard threaded bottle tops
- Designed for use on glass or polystyrene media bottles
- Attaches to most popular 33 and 45 mm Ø bottle's necks with standard GPI threads
Units are color coded by membrane type (CA: Orange; NYL: Red, CN: Blue; and PES: Yellow). Choice of membrane media: Cellulose acetate (CA), for low protein binding. Nylon (NYL), for low extractable. Cellulose Nitrate (CN), for general purpose filtration. Or polyethersulfone (PES), for fast flow rates and low protein binding.
Membranes come in 0.20, 0.22 or 0.45 µm pore sizes. All four-type membranes are detergent-free, tissue culture compatible, and heat-sealed to the support grid to maximize flow rate, reduce foaming and protein denaturization.