Micro Cover Glasses, Square and Rectangular Sizes, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
The Micro Cover Glasses are manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant.
- Uniform in size and thickness
- Square slides are 22 x 22 mm
- Rectangular slides are 22 mm wide and 30, 40 or 50 mm long
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.