Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
This pen helps to flatten and separate tissue sections and reduces compression in thin sections.
- Powered by AA batteries
Delivery: Available in two models: Wax Pen 1 is powered by one AA battery; Wax Pen 2 is powered by two AA batteries.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.