Spécifications
- Description:Hardened Steel Deburring Tool Set
- Supplier No.:62161-SD
Spécifications
A propos de cet article
Made from hardened steel, this tool has three razor sharp cutting edges ranging from a very sharp point to one that is approximately 10 mm in length.
These nickel plated triangular tools are mounted in red and black plastic handles. Tool length is 30 mm by 2.1 mm across (1.2×0.08"). The overall length of the tool is 154 mm (5.7"). Two tools/set.