Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
- Made from a special plastic that minimizes static
- Complete with grid recording card
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.