About this item
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection.
- Economical
- Ambidextrous style
- Beaded cuff
Chemically resistant to acids, alcohols, ketones and alkalis. Non-sterile.
Available either with powdered (biologically cornstarch dusting, ~2 mg/glove) or powder-free gloves.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102097-390
- 102097-404
- 102097-406
- 102097-388
- 102097-396
- 102097-398
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free