Spécifications
- Description:PSA no hole
- Supplier No.:50318-05
Spécifications
A propos de cet article
This flocked cloth is made by binding a rayon flock to a woven cotton backing; it is the most popular in our line.
- Available with Pressure Sensitive Adhesive (PSA) with and without hole
It is excellent for Final polishing when used in conjunction with diamond or alumina polishing compounds.