Frisket Knife, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
The frisket Knife is lightweight and disposable. Designed for easy handling.
Comes with a fixed #11 scalpel blade, and a locking safety cap when it is not in use.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.