Flat Emery Sticks, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Emery paper which is glued to a wooden handle. Very useful for filing excess resin of sample block tips. Grit size ranges from number 4/0 (finest) to 3 (coarsest).
Dimensions L x W: 279 x 19.1 mm (11 x 3/4").
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.