Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
A plastic handle, 267 mm (10 1/2") long, holds strips of emery paper or cloth 279 x 25.4 mm (11 x 1"). Strips are easily replaced. Comes complete with 2 strips/set (grit 120 and 240).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.