3 Cavities Spot Plate, LDPE, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
These spot plates have three depressions measuring 21 mm in diameter and 7 mm in depth.
- Made from low density polyethylene
- Withstands temperatures up to 80 °C
Tray dimensions: 28×85 mm
Shipping weight: 0.6 lbs. per case of 40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.