12 Cavities Spot Plate, Polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Very similar to the white porcelain plate, this PP plate comprises 12 cavities of approximately 1 ml capacity and is economically priced. This plate is very high quality.
- Unbreakable
- Autoclavable
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.