A newly designed mold which is convenient and offers better embedments. This tray-and-mold in one unit offers a unique specimen block that no other mold can. Each tray contains 2×10 block molds and measures 87×66 mm.
Available in two sizes:
Size 5.6 mm diameter, which corresponds to gelatin and BEEM® capsules size 3, and forms a block face of 1.6 mm.
Size 8 mm diameter, which corresponds to gelatin and BEEM® capsule size ;00' and forms a block face of 3 mm.