Dust-Off® Microfiber Cleaning Cloth, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
For superior cleaning of delicate surfaces.
- Environmentally responsible - lifts away oily film and dirt with no chemicals
- Will not streak or smear surfaces
- Non-abrasive and 100% lint-free
- Reusable - machine or hand washable - long lasting
- Size: 7½×7½" (19×19 cm)
Ideal for camera, lenses/filters, microscopes, computer, eyeglasses, plastic, metal, or any smooth surface.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.