DuraSeal™ Polyethylene Stretch Film, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
This Durable heat-resistant, solvent-proof laboratory stretch film forms an airtight heat-resistant barrier.
- Microwave compatible and tear resistant
- Chemically resistant to all solvents and caustic agents
- Withstands a temperature of 100 °C
- Able to stretch 500 to 800% before breaking or tearing
- Able to cling to self following stretching
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.