Dispensing Bottles, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Low density polyethylene bottles.
- Tip has a 0.76 mm (0.003") opening
By squeezing the bottle a steady stream of liquid is produced.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.