Diamond Knife Cleaning Kit, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
This cleaning kit is used for the removal of sections from the knife edge and the trough.
Hair curl is made from human hair; good for removing sections, and cleaning the diamond knife.
Delivery: Kit includes 5×5 ml vials of cleaning solution, 4 rods for cleaning the knife edge, and 1 hair curl.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.