Steam Autoclaves Imprinted Tape, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Indicate the status of all laboratory items to be sterilized with these tape labels.
The word 'STERILIZED' appears after processing at 250 °F under normal sterilization conditions. Has 3" core.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.