Spécifications
- Description:Cutting Board
- Material:PP
- Supplier No.:63308
Spécifications
A propos de cet article
This cutting board is autoclavable, unbreakable, non-absorbent, and easy to clean
Made in the US of NSF, FDA and USDA approved material
Measurements L x W x H: 300 x 210 x 9 mm
It is a useful item in any laboratory. Our cutting boards are made from pure Polypropylene which can be autoclaved repeatedly. Polypropylene can withstand temperatures up to 135 Degrees F. Polypropylene can be sterilized via autoclave, microwave, gas sterilization, chemical disinfectant sterilization.