Specifications
- Description:Cryo sleeve
- Length:27.5 cm
- Cat. no.:100490-098
- Supplier No.:61082-10
Specifications
About this item
Clear polyvinyl chloride. Ideal for enclosing Cryo Storage Canes for extra security during handling and storage. Fits over most other canes. Will not become brittle during freezing or thawing. Will not shred or tear after repeated use like cardboard. Length: 27.5 cm (10 13/16").